siRNA Details
| Gene Symbol: Rcbtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033185.1 | 1, 2 | 409 | |||
| GenBank mRNA | AK013119.1 | - | 1, 2 | 485 | ||
| AK013540.1 | - | 189 | ||||
| AK147784.1 | - | 1, 2 | 30 | |||
| AK160881.1 | - | 1, 2 | 131 | |||
| BB854398.1 | - | 1, 2 | 410 | |||

Rcbtb2
Genomic Map