siRNA Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029425.1 | 10 | 2296 | |||
| GenBank mRNA | AJ237846.1 | - | 10 | 2226 | ||
| AK004162.1 | - | 1 | 372 | |||
| AK087940.1 | - | 4 | 1401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbmx| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029425.1 | 10 | 2296 | |||
| GenBank mRNA | AJ237846.1 | - | 10 | 2226 | ||
| AK004162.1 | - | 1 | 372 | |||
| AK087940.1 | - | 4 | 1401 | |||