siRNA Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029425.1 | 10 | 2175 | |||
| GenBank mRNA | AJ237846.1 | - | 10 | 2104 | ||
| AK004162.1 | - | 1 | 251 | |||
| AK008618.1 | - | 1 | 710 | |||
| AK087940.1 | - | 4 | 1280 | |||
| AK165006.1 | - | 10 | 2122 | |||

Rbmx
Genomic Map