siRNA Details
| Gene Symbol: A530053G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015565.2 | 2 | 864 | |||
| GenBank mRNA | AK080087.1 | - | 2 | 865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A530053G22Rik| Gene Symbol: A530053G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015565.2 | 2 | 864 | |||
| GenBank mRNA | AK080087.1 | - | 2 | 865 | ||