siRNA Details
| Gene Symbol: 4933427E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033197.1 | 1 | 793 | |||
| GenBank mRNA | AK006089.1 | - | 1 | 607 | ||
| AK006196.1 | - | 1 | 582 | |||
| AK006985.1 | - | 1 | 604 | |||
| AK016947.1 | - | 1 | 794 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933427E11Rik| Gene Symbol: 4933427E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033197.1 | 1 | 793 | |||
| GenBank mRNA | AK006089.1 | - | 1 | 607 | ||
| AK006196.1 | - | 1 | 582 | |||
| AK006985.1 | - | 1 | 604 | |||
| AK016947.1 | - | 1 | 794 | |||