siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027869.1 | 18, 19 | 2082 | |||
| NR_027870.1 | 17, 18 | 2061 | ||||
| GenBank mRNA | AJ003007.1 | - | 18, 19 | 1950 | ||
| AK019068.1 | - | 1, 2 | 37 | |||
| AK043436.1 | - | 17, 18 | 2019 | |||

Dtnb
Genomic Map