siRNA Details
| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027869.1 | 19 | 2103 | |||
| NR_027870.1 | 18 | 2082 | ||||
| GenBank mRNA | AJ003007.1 | - | 19 | 1971 | ||
| AK019068.1 | - | 2 | 58 | |||
| AK043436.1 | - | 18 | 2040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtnb| Gene Symbol: Dtnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027869.1 | 19 | 2103 | |||
| NR_027870.1 | 18 | 2082 | ||||
| GenBank mRNA | AJ003007.1 | - | 19 | 1971 | ||
| AK019068.1 | - | 2 | 58 | |||
| AK043436.1 | - | 18 | 2040 | |||