siRNA Details
| Gene Symbol: Zp4-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027813.1 | 5 | 672 | |||
| GenBank mRNA | DQ279480.1 | - | 5 | 672 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV655471.1 | - | 5 | 570 | Chr. 13 - 11522154 - 11526436 [+] on Build GRCm38 | |

Zp4-ps
Genomic Map