siRNA Details
| Gene Symbol: 4933432I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015575.1 | 3 | 297 | |||
| NR_027843.1 | 3 | 442 | ||||
| GenBank mRNA | AK005656.1 | - | 3 | 442 | ||
| AK016380.1 | - | 2 | 289 | |||
| AK017024.1 | - | 3 | 298 | |||
| AK161480.1 | - | 443 | ||||

4933432I09Rik
Genomic Map