siRNA Details
| Gene Symbol: 4933432I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015575.1 | 3 | 536 | |||
| NR_027843.1 | 3 | 681 | ||||
| GenBank mRNA | AK016380.1 | - | 2 | 528 | ||
| AK017024.1 | - | 3 | 537 | |||
| AK161480.1 | - | 682 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933432I09Rik| Gene Symbol: 4933432I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015575.1 | 3 | 536 | |||
| NR_027843.1 | 3 | 681 | ||||
| GenBank mRNA | AK016380.1 | - | 2 | 528 | ||
| AK017024.1 | - | 3 | 537 | |||
| AK161480.1 | - | 682 | ||||