siRNA Details
| Gene Symbol: 4933432I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015575.1 | 3 | 1028 | |||
| NR_027843.1 | 3 | 1173 | ||||
| GenBank mRNA | AK017024.1 | - | 3 | 1029 | ||
| AK161480.1 | - | 1174 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933432I09Rik| Gene Symbol: 4933432I09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015575.1 | 3 | 1028 | |||
| NR_027843.1 | 3 | 1173 | ||||
| GenBank mRNA | AK017024.1 | - | 3 | 1029 | ||
| AK161480.1 | - | 1174 | ||||