siRNA Details
| Gene Symbol: Dnm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002870.2 | 1 | 3282 | |||
| GenBank mRNA | AK053654.1 | - | 1 | 2066 | ||
| AK164873.1 | - | 1 | 1110 | |||
| AY751519.1 | - | 1 | 3242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnm3os| Gene Symbol: Dnm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002870.2 | 1 | 3282 | |||
| GenBank mRNA | AK053654.1 | - | 1 | 2066 | ||
| AK164873.1 | - | 1 | 1110 | |||
| AY751519.1 | - | 1 | 3242 | |||