siRNA Details
Gene Symbol: Phtf1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030676.1 | 1 | 1637 | |||
GenBank mRNA | AK048683.1 | - | 1 | 1638 | ||
AK051503.1 | - | 1 | 1635 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Phtf1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030676.1 | 1 | 1637 | |||
GenBank mRNA | AK048683.1 | - | 1 | 1638 | ||
AK051503.1 | - | 1 | 1635 |