siRNA Details
| Gene Symbol: Phtf1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030676.1 | 1 | 3105 | |||
| GenBank mRNA | AK048683.1 | - | 1 | 3106 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phtf1os| Gene Symbol: Phtf1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030676.1 | 1 | 3105 | |||
| GenBank mRNA | AK048683.1 | - | 1 | 3106 | ||