siRNA Details
| Gene Symbol: Gm10649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028579.1 | 2 | 806 | |||
| GenBank mRNA | AK138650.1 | - | 2 | 807 | ||
| AK162930.1 | - | 2 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10649| Gene Symbol: Gm10649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028579.1 | 2 | 806 | |||
| GenBank mRNA | AK138650.1 | - | 2 | 807 | ||
| AK162930.1 | - | 2 | 807 | |||