siRNA Details
| Gene Symbol: Gm10409 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033121.1 | 7 | 1397 | |||
| Gene Symbol: Gm3020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK012594.1 | - | 7 | 1400 | ||

Gm10409
Genomic Map