siRNA Details
| Gene Symbol: Gm1141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027801.1 | 9 | 2087 | |||
| GenBank mRNA | AK133067.1 | - | 9 | 2088 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1141| Gene Symbol: Gm1141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027801.1 | 9 | 2087 | |||
| GenBank mRNA | AK133067.1 | - | 9 | 2088 | ||