siRNA Details
| Gene Symbol: Nlrp1c-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027858.1 | 8 | 2530 | |||
| GenBank mRNA | BC141384.1 | - | 8 | 2526 | 9055997 | |
| DQ117602.1 | - | 8 | 2530 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp1c-ps| Gene Symbol: Nlrp1c-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027858.1 | 8 | 2530 | |||
| GenBank mRNA | BC141384.1 | - | 8 | 2526 | 9055997 | |
| DQ117602.1 | - | 8 | 2530 | |||