siRNA Details
| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027644.1 | 1 | 52 | |||
| GenBank mRNA | BC064106.1 | - | 1 | 52 | 1528324 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vsig8| Gene Symbol: Vsig8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027644.1 | 1 | 52 | |||
| GenBank mRNA | BC064106.1 | - | 1 | 52 | 1528324 | |