siRNA Details
| Gene Symbol: Gm2518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015538.1 | 1 | 1145 | |||
| GenBank mRNA | AK041085.1 | - | 2 | 1041 | ||
| AK077736.1 | - | 1 | 1142 | |||
| AK082962.1 | - | 1 | 1144 | |||
| AK085400.1 | - | 1 | 1117 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2518| Gene Symbol: Gm2518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015538.1 | 1 | 1145 | |||
| GenBank mRNA | AK041085.1 | - | 2 | 1041 | ||
| AK077736.1 | - | 1 | 1142 | |||
| AK082962.1 | - | 1 | 1144 | |||
| AK085400.1 | - | 1 | 1117 | |||