siRNA Details
| Gene Symbol: Gm2518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015538.1 | 1 | 1537 | |||
| GenBank mRNA | AK077736.1 | - | 1 | 1534 | ||
| AK082962.1 | - | 1 | 1536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm2518| Gene Symbol: Gm2518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015538.1 | 1 | 1537 | |||
| GenBank mRNA | AK077736.1 | - | 1 | 1534 | ||
| AK082962.1 | - | 1 | 1536 | |||