siRNA Details
Gene Symbol: 4930503E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028310.1 | 4 | 1318 | |||
GenBank mRNA | AK015688.1 | - | 4 | 1319 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930503E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028310.1 | 4 | 1318 | |||
GenBank mRNA | AK015688.1 | - | 4 | 1319 |