siRNA Details
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003568.1 | 7 | 1618 | |||
GenBank mRNA | AF154337.1 | - | 7 | 1602 | ||
AK170103.1 | - | 7 | 1612 | |||
AK171335.1 | - | 7 | 1612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003568.1 | 7 | 1618 | |||
GenBank mRNA | AF154337.1 | - | 7 | 1602 | ||
AK170103.1 | - | 7 | 1612 | |||
AK171335.1 | - | 7 | 1612 |