siRNA Details
| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003568.1 | 7 | 2198 | |||
| GenBank mRNA | DV659289.1 | - | 1 | 491 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU058094.1 | - | 1 | 57 | Chr. 13 - 12614166 - 12614864 [-] on Build GRCm38 | |

Gpr137b-ps
Genomic Map