siRNA Details
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003568.1 | 7 | 2477 | |||
GenBank mRNA | CN701965.1 | - | 1 | 111 | ||
DV659289.1 | - | 1 | 770 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpr137b-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003568.1 | 7 | 2477 | |||
GenBank mRNA | CN701965.1 | - | 1 | 111 | ||
DV659289.1 | - | 1 | 770 |