siRNA Details
| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003568.1 | 7 | 2511 | |||
| GenBank mRNA | CN701965.1 | - | 1 | 145 | ||
| DV659289.1 | - | 1 | 804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr137b-ps| Gene Symbol: Gpr137b-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003568.1 | 7 | 2511 | |||
| GenBank mRNA | CN701965.1 | - | 1 | 145 | ||
| DV659289.1 | - | 1 | 804 | |||