siRNA Details
| Gene Symbol: D730001G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027836.1 | 3 | 662 | |||
| NR_027837.1 | 2 | 563 | ||||
| GenBank mRNA | AK021322.1 | - | 2 | 564 | ||
| AK037167.1 | - | 2 | 563 | |||
| BC055822.1 | - | 3 | 627 | |||
| BC147771.1 | - | 1 | 443 | |||
| BC147774.1 | - | 1 | 443 | |||

D730001G18Rik
Genomic Map