siRNA Details
| Gene Symbol: D730001G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027836.1 | 3 | 1570 | |||
| NR_027837.1 | 2 | 1471 | ||||
| GenBank mRNA | AK037167.1 | - | 2 | 1471 | ||
| BC055822.1 | - | 3 | 1535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730001G18Rik| Gene Symbol: D730001G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027836.1 | 3 | 1570 | |||
| NR_027837.1 | 2 | 1471 | ||||
| GenBank mRNA | AK037167.1 | - | 2 | 1471 | ||
| BC055822.1 | - | 3 | 1535 | |||