siRNA Details
| Gene Symbol: Pnpla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028142.1 | 2, 3 | 363 | |||
| GenBank mRNA | BC019188.1 | - | 2, 3 | 345 | 5027544 | |
| BC044737.1 | - | 2, 3 | 327 | |||
| BC044781.2 | - | 2, 3 | 327 | 5151911 | ||
| BC054825.1 | - | 2, 3 | 322 | |||

Pnpla2
Genomic Map