siRNA Details
| Gene Symbol: Prmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024139.2 | 2 | 2569 | |||
| GenBank mRNA | AK087551.1 | - | 2 | 2117 | ||
| AK157132.1 | - | 3 | 2523 | |||
| BC066221.1 | - | 2 | 2546 | 30436952 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt6| Gene Symbol: Prmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024139.2 | 2 | 2569 | |||
| GenBank mRNA | AK087551.1 | - | 2 | 2117 | ||
| AK157132.1 | - | 3 | 2523 | |||
| BC066221.1 | - | 2 | 2546 | 30436952 | ||