siRNA Details
| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028056.1 | 3 | 267 | |||
| GenBank mRNA | AK216483.1 | - | 2 | 38 | ||
| CV562852.1 | - | 3 | 206 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rit1| Gene Symbol: Rit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028056.1 | 3 | 267 | |||
| GenBank mRNA | AK216483.1 | - | 2 | 38 | ||
| CV562852.1 | - | 3 | 206 | |||