siRNA Details
| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024025.1 | 3 | 385 | |||
| GenBank mRNA | AK133889.1 | - | 1 | 47 | ||
| BC063050.1 | - | 3 | 369 | 5716297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr2| Gene Symbol: Mtmr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024025.1 | 3 | 385 | |||
| GenBank mRNA | AK133889.1 | - | 1 | 47 | ||
| BC063050.1 | - | 3 | 369 | 5716297 | ||