siRNA Details
| Gene Symbol: Gm10638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027829.1 | 1 | 1318 | |||
| GenBank mRNA | AK133595.1 | - | 844 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10638| Gene Symbol: Gm10638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027829.1 | 1 | 1318 | |||
| GenBank mRNA | AK133595.1 | - | 844 | |||