siRNA Details
| Gene Symbol: Rprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024198.2 | 1 | 42 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rprl3| Gene Symbol: Rprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024198.2 | 1 | 42 | |||