siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 5 | 605 | |||
| GenBank mRNA | AK043630.1 | - | 5 | 606 | ||
| AK140288.1 | - | 5 | 606 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY727408.1 | - | 5 | 595 | Chr. 7 - 55962541 - 55976908 [+] on Build GRCm38 | |

A230056P14Rik
Genomic Map