siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 6 | 1373 | |||
| GenBank mRNA | AK043630.1 | - | 6 | 1374 | ||
| AK075669.1 | - | 717 | ||||
| AK140288.1 | - | 6 | 1375 | |||
| BC147226.1 | - | 4 | 1012 | |||
| BC147227.1 | - | 4 | 1012 | |||

A230056P14Rik
Genomic Map