siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 1 | 83 | |||
| GenBank mRNA | AK038718.1 | - | 1 | 84 | ||
| AK039340.1 | - | 1 | 79 | |||
| AK043630.1 | - | 1 | 84 | |||
| AK140288.1 | - | 1 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230056P14Rik| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 1 | 83 | |||
| GenBank mRNA | AK038718.1 | - | 1 | 84 | ||
| AK039340.1 | - | 1 | 79 | |||
| AK043630.1 | - | 1 | 84 | |||
| AK140288.1 | - | 1 | 84 | |||