siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 4 | 508 | |||
| GenBank mRNA | AK043630.1 | - | 4 | 509 | ||
| AK140288.1 | - | 4 | 509 | |||
| BC147226.1 | - | 2 | 147 | |||
| BC147227.1 | - | 2 | 147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230056P14Rik| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 4 | 508 | |||
| GenBank mRNA | AK043630.1 | - | 4 | 509 | ||
| AK140288.1 | - | 4 | 509 | |||
| BC147226.1 | - | 2 | 147 | |||
| BC147227.1 | - | 2 | 147 | |||