siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015495.2 | 6 | 1034 | |||
| GenBank mRNA | AK043630.1 | - | 6 | 1035 | ||
| AK075669.1 | - | 377 | ||||
| AK140288.1 | - | 6 | 1036 | |||
| BC147226.1 | - | 4 | 673 | |||
| BC147227.1 | - | 4 | 673 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB609724.1 | - | 2 | 377 | Chr. 7 - 55976903 - 55980004 [+] on Build GRCm38 | |

A230056P14Rik
Genomic Map