siRNA Details
| Gene Symbol: Rn45s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046233.2 | 4037 | ||||
| Gene Symbol: Rn18s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003278.3 | 30 | ||||
| GenBank mRNA | AK135936.1 | - | 30 | |||
| AK157747.1 | - | 30 | ||||
| AK171941.1 | - | 30 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NR_046237, NR_046239, FQ227804, FQ227006, FQ234841, and others.

Rn45s
Genomic Map