siRNA Details
| Gene Symbol: 2900052N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015605.1 | 2 | 315 | |||
| GenBank mRNA | AK038437.1 | - | 2 | 316 | ||
| AK049847.1 | - | 2 | 315 | |||
| AK140089.1 | - | 2 | 288 | |||
| BC054400.1 | - | 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900052N01Rik| Gene Symbol: 2900052N01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015605.1 | 2 | 315 | |||
| GenBank mRNA | AK038437.1 | - | 2 | 316 | ||
| AK049847.1 | - | 2 | 315 | |||
| AK140089.1 | - | 2 | 288 | |||
| BC054400.1 | - | 2 | 291 | |||