siRNA Details
| Gene Symbol: Zfp783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027963.1 | 6 | 4337 | |||
| GenBank mRNA | AK220535.1 | - | 6 | 4209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp783| Gene Symbol: Zfp783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027963.1 | 6 | 4337 | |||
| GenBank mRNA | AK220535.1 | - | 6 | 4209 | ||