siRNA Details
| Gene Symbol: Zfp783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027963.1 | 6 | 2992 | |||
| GenBank mRNA | AK039184.1 | - | 6 | 2560 | ||
| AK220535.1 | - | 6 | 2864 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp783| Gene Symbol: Zfp783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027963.1 | 6 | 2992 | |||
| GenBank mRNA | AK039184.1 | - | 6 | 2560 | ||
| AK220535.1 | - | 6 | 2864 | |||