siRNA Details
| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015606.3 | 2 | 1049 | |||
| GenBank mRNA | AK035234.1 | - | 1 | 1558 | ||
| AK035285.1 | - | 2 | 1180 | |||
| AK051953.1 | - | 2 | 927 | |||
| AK161046.1 | - | 2 | 1051 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r12b| Gene Symbol: Ppp1r12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015606.3 | 2 | 1049 | |||
| GenBank mRNA | AK035234.1 | - | 1 | 1558 | ||
| AK035285.1 | - | 2 | 1180 | |||
| AK051953.1 | - | 2 | 927 | |||
| AK161046.1 | - | 2 | 1051 | |||