siRNA Details
| Gene Symbol: Mgat4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145149.1 | - | 1, 2 | 71 | 9053700 | |
| BC145786.1 | - | 1, 2 | 71 | 40131089 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4e| Gene Symbol: Mgat4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC145149.1 | - | 1, 2 | 71 | 9053700 | |
| BC145786.1 | - | 1, 2 | 71 | 40131089 | ||