siRNA Details
| Gene Symbol: Gm10941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026944.1 | 1 | 245 | |||
| GenBank mRNA | AK132904.1 | - | 1 | 246 | ||
| AK164976.1 | - | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10941| Gene Symbol: Gm10941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026944.1 | 1 | 245 | |||
| GenBank mRNA | AK132904.1 | - | 1 | 246 | ||
| AK164976.1 | - | 1 | 95 | |||