siRNA Details
| Gene Symbol: Dlx1as | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002854.2 | 2 | 2613 | |||
| GenBank mRNA | AK132348.1 | - | 2 | 2614 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx1as| Gene Symbol: Dlx1as | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002854.2 | 2 | 2613 | |||
| GenBank mRNA | AK132348.1 | - | 2 | 2614 | ||