siRNA Details
| Gene Symbol: 1500015A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029432.1 | 1 | 1102 | |||
| GenBank mRNA | AK005240.1 | - | 1 | 1102 | ||
| AK133612.1 | - | 1 | 1086 | |||
| BC047208.1 | - | 1 | 1037 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU504362.1 | - | 1 | 582 | Chr. 18 - 61726911 - 61727801 [+] on Build GRCm38 | |

1500015A07Rik
Genomic Map