siRNA Details
| Gene Symbol: 1500015A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029432.1 | 1 | 618 | |||
| GenBank mRNA | AK005240.1 | - | 1 | 618 | ||
| AK133612.1 | - | 1 | 602 | |||
| BC047208.1 | - | 1 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500015A07Rik| Gene Symbol: 1500015A07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029432.1 | 1 | 618 | |||
| GenBank mRNA | AK005240.1 | - | 1 | 618 | ||
| AK133612.1 | - | 1 | 602 | |||
| BC047208.1 | - | 1 | 553 | |||