siRNA Details
Gene Symbol: 4933406K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015512.2 | 8 | 1056 | |||
GenBank mRNA | AK015208.1 | - | 8 | 1057 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933406K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015512.2 | 8 | 1056 | |||
GenBank mRNA | AK015208.1 | - | 8 | 1057 |